Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avidin (from Egg white)

Supplier: MP Biomedicals
0215004780 0215004725 0215004710
76204-018EA 304.05 USD InStock
76204-018 76204-016 76204-020
Avidin (from Egg white)
Proteins and Peptides
Avidin is a basic glycoprotein isolated from raw egg white. It exhibits high binding affinity for biotin and is capable of producing biotin deficiency in rats and chicks. It also occurs in the white portion of eggs and the oviducts of birds and amphibia. It is destroyed by cooking or irradiation.

  • Ready for CE IVD labeling of clinical applications
  • Specific activity: 10 to 15 u/mg solid
  • Solubility: Soluble in water or dilute aqueous buffer
  • Unit definition: One unit will bind 1 µg of D-biotin at pH 8.9

Avidin is useful in a biotin-avidin system for labeling biomolecules for receptor studies, immunoassays and immunohistological methods. Avidin egg white was used in an assay using functionalized xenon as a biosensor to detect biotin-avidin binding. Egg white was used at 80 nmol. Egg white avidin has been used as a cross-linker for biotin-conjugated antibodies. Avidin also been used as a blocking agent in ELISA applications.

Caution: Research use only (RUO).

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty