Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Yeast Extract, Yellow-Tan Powder for Molecular Biology, MP Biomedicals

Supplier: MP Biomedicals
0219402783 0219402791
76204-082EA 86.6 USD InStock
76204-082 76204-084
Yeast Extract, Yellow-Tan Powder for Molecular Biology, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Yeast extract powder is a vacuum dried autolyzed yeast extract produced from 100% Baker's yeast. It is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.

  • Grade : Molecular Biology Reagent
  • Suitability : Suitable for alternative to beef extract
  • Key Applications : Bacteriological media component
  • Application Areas : Used as a bacteriological media component
  • Product Type : Proteins, Enzymes & Peptides
  • Presentation : Yellow-tan Powder
  • pH : 5 - 5.5 (aqueous soln.)(Lit.)
  • Foreign activity : Salmonella - Negative
  • Moisture content : <5%
  • NOTES : Mold Count ≤50/g, Standard Plate Count ≤50,000/g
  • Solubility : Solubility in water: - Clear (5 g/100 mL).
  • Mold Count: < 50/gram
  • Standard Plate Count: < 50,000/gram
  • Salmonella: Negative

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty