Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger
Synonyms: Muriatic acid

0219405480 0219405490 0219405491
76204-102EA 107.97 USD InStock
76204-102 76204-098 76204-100
Hydrochloric acid ACS
Hydrochloric acid
  • Grade ACS Reagent Grade
  • Assay 36.5 - 38%
  • Product Type Inorganics
  • Biochemical Category Acids
  • Vapor Density 1.267 (air = 1)(Lit.)
  • Vapor Pressure 3.23 psi ( 21.1 °C) (Lit.); 7.93 psi ( 37.7 °C) (Lit.)
  • Presentation Clear, Colorless Liquid
  • Color (APHA) ≤10
  • Heavy Metals Lead ≤1 ppm
  • Residue on ignition ≤5 ppm
  • NOTES Bromide (Br) ≤0.005%
  • Sulfate (SO4) ≤1 ppm
  • Sulfite (SO3) ≤1 ppm
  • Free Chlorine (Cl) ≤1 ppm
  • Ammonium (NH4) ≤3 ppm
  • Arsenic (As) ≤0.01 ppm
  • Iron (Fe) ≤0.2 ppm
  • Extractable Organic Substances: ≤5 ppm


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: >100 °C (lit.)
Melting Pt: –114.4 °C (lit.)
Density: 1.161...1.19 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: ,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/41/15/36384115.jpg
Order Now
Specification Test Results
Assay 36.5 - 38%
Appearance Free from suspended matter or sediment,
Color (APHA) 10
Residue after Ignition 5 ppm
Bromide (Br) 0.005%
Sulfate (SO4) 1 ppm
Free Chlorine (CI) 1 ppm
Ammonium (NH4) 3 ppm
Arsenic (As) 0.01 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty