Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine Aprotinin (from Lung)

Supplier: MP Biomedicals
0219410180 0219410125 0219410105
76204-328EA 104.84 USD InStock
76204-328 76204-332 76204-330
Bovine Aprotinin (from Lung)
Enzymes
Aprotinin is found in bovine lymph nodes, lung, parotid gland, spleen, liver, pancreas, seminal vesicles, thyroid gland, kidney, mucous membranes of the trachea and esophagus, ovaries, heart, posterior pituitary and cartilage. It is a serine protease inhibitor which is reactive against trypsin, chymotrypsin, plasmatic and glandular kininogenases, plasmin, kallkrein, urokinase, clotting factor XIIa, protein C, proteinases of the complement system, leukocyte and tissue proteinases. It does not inhibit thrombin. Aprotinin works by blocking the active sites of enzymes. Binding is reversible with most aprotinin-protease complexes dissociating at pH > 10 or < 3.

  • Specific Activity : ≥3 TIU/mg
  • Key Applications : Inhibitor Stain
  • Application Areas : Molecular biology
  • Product Type : Biochemicals
  • Biochemical Category : Pharmaceuticals
  • Extinction Coefficient (E1%) : (280 nm) = 8.3 (water)
  • UV/Visible Absorbance : λ max (water) - 277 ± 5 nm
  • Presentation : White Powder
  • White Powder : 10.5
  • pH : 5.0 - 7.0 (1% aq soln)
  • Water Content : ≤6%
  • Solubility : Soluble in water (10 mg/mL - completely soluble) and in aqueous buffers of low ionic strengths.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty