Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Standard Line Sterile Centrifuge Tubes with Flat or Plug Caps, PP

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
CTNA301 CTNA314 CTNA214 CTNA305 CTNA312 CTNA211 CTNA311 CTNA209 CFT115500 CTNA213 CTNA216 CTNA207 CTNA210 CTNA303 CFT114500 CTNA208 CTNA313 CFT113500 CTNA215 CTNA304
10026-078CS 224.37 USD InStock
10026-078 76204-402 76204-404 10026-074 75845-202 10026-076 75845-206 10025-702 75845-204 10025-712 10025-700 10025-688 10025-698 10025-686 10025-696 10025-694 76204-542 10025-692 76204-400 10025-690
VWR® Standard Line Sterile Centrifuge Tubes with Flat or Plug Caps, PP
Tubes Centrifuge Tubes
These disposable conical-bottom and freestanding tubes are manufactured from high-quality USP Class 6 gamma-resistant polypropylene.

  • Available in two volumes: 15 and 50 ml
  • Available with two bottom styles: conical bottom and self-standing bottom
  • Longer length screw caps with sealing ring prevent leakage
  • Easy-to-read black graduations are accurate to ±2%, with 1 ml increments (15 ml) or 2.5 ml increments (50 ml)
  • Large, unerasable frosted white writing area
  • Both the graduations and writing areas are chloroform-resistant
  • Engraved graduation at the conical bottom of each tube and leak-proof
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters

They feature black printed graduations and smooth inner walls for easy filling and sample preparation. Tubes are RNase-free, DNase-free, and non-pyrogenic.

Caps are available in two styles: flat, with a smooth surface for additional labeling options, and plug style for a secure seal when using shakers and horizontal incubators. All caps are made of white high-density polyethylene.

A white printed writing area on the side allows for permanent coding. Tubes and caps are autoclavable at 121 °C and freezable at –80 °C.

Max. rotation speed up to 12500×g for conical bottom tubes, and 10000×g for self-standing tubes.

Certifications: ISO 13485:2003 and ISO 9001:2008.

Ordering information: All tubes are packaged with caps attached and sterilized with irradiation. Plastic racks are available as optional accessories in seven colors. An assorted color pack offers one of each color: blue, green, orange, red, and yellow. Racks are designed for use with plug or flat bottom tubes. Cap and tube packed separately for items 75845-202, 75845-204, and 75845-206.

Packaging: Racked tubes are packaged in environmentally responsible (fully recyclable) chipboard racks or plastic racks. Bagged tubes are packaged in durable and re-sealable (zip closure) bags. Inner packaging is individually labelled for lot-to-lot traceability.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty