Would you like to log in again?
DuoClean™ Wipes, Berkshire
Supplier: Berkshire
DCVPBL0512 DCVPHP3012 DCVPHP6012
76204-556CS 692.32 USD InStock
76204-556 76204-558 76204-560
DuoClean™ Wipes, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
DuoClean™ wipes provide convenience, consistency, disinfectant and sterilant capabilities for cleaning a wide array of cleanroom surfaces.
  • Lightweight
  • Burstable pouch wet-to-dry technology
  • MicroSeal-VP 100% continuous filament polyester knit
  • Ultrasonically sealed edge for reduced fiber contamination
  • Laundered and packaged in ISO Class 4 cleanroom
The advanced cleaning system supports the cleanroom operator in a scalable session format.
Certifications: ISO Class 3-7.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty