Would you like to log in again?
Rotors for Centrifuge 5425/5425 R
Supplier: Eppendorf
5495503005 5495512004 5495512004 5495500006 5495508007 5495500006 5495510001 5495510001 5495508007 5495505008 5495503005 5495505008
76204-616EA 1505.84 USD InStock
CA76204-626 76204-616 76204-626 CA76204-616 CA76204-624 76204-628 CA76204-614 76204-612 CA76204-612 76204-614 76204-624 CA76204-628
Rotors for Centrifuge 5425/5425 R
Centrifuges Rotors
Eppendorf rotors for centrifuge 5425/5425 R, precision meets versatility. Optimize your centrifuge with a range of capacities and speeds. Explore accessory options such as adaptors and buckets for tailored sample processing, ensuring optimal results in molecular biology and beyond.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight and PTFE coated options available
Refine your centrifugation experience with Eppendorf rotors designed for centrifuge 5425/5425 R. Offering versatility in capacities and speeds, these rotors cater to a spectrum of applications, from microcentrifugation to larger sample volumes. Accessories such as adaptors and buckets further customize your setup for specific needs. Achieve reliable sample separation with precision and flexibility, meeting the demands of molecular biology, genomics, and beyond. Explore the comprehensive range to optimize your centrifuge for reproducible results.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty