Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ReadyStrain™ Sterile Cell Straining Kits, MTC Bio
Supplier: MTC Bio
C5100 C5070
76204-644PK 226.96 USD InStock
76204-644 76204-646
ReadyStrain™ Sterile Cell Straining Kits, MTC Bio
Cell Strainers
ReadyStrain™ is the first product of its kind, providing all the necessary parts for filtration of cell suspensions and combining them into a single, pre-assembled, ready to use, sterile packet.
  • Pre-assembled & supplied ready to use
  • Opens directly under the hood to reduce contamination risks
  • Garaunteed sterile via gamma sterilization
  • Advanced design provides faster flow-through
These kits eliminate the contamination risks associated with opening multiple packages, and assembling separate pieces (i.e. tubes, strainers, and caps). The individual sterile packets mean researchers are no longer forced to expose entire bags of 50 mL tubes and corresponding caps in order to perform one filtration. Instead they can open a single ReadyStrain™ packet directly under the hood. Processing and storage after the filtration is complete.
Delivery information: Pre-assembled sterile cell straining kits include a premium cell strainer, a 50 mL tube, and separate cap.
Packaging: Packaged in easy-open "tear-strip" bags.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty