Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PREVI® Color Gram, BioMerieux
Supplier: bioMérieux
414292
76204-648EA 32817.5 USD InStock
76204-648
PREVI® Color Gram, BioMerieux
Stains Microbiology Stains
Fully automated stain process from fixation to drying.
Includes PREVI® Color Gram Accessory Kit and PREVI® Color Gram Main Kit:
PREVI® Color Gram Accessory Kit (Contains 12 Slide Carosel, Waste Tubing, Waste Container V2, and Maintenance Kit V2)
PREVI® Color Gram Main Kit (Contains Priming Tool, Silicon Grease, Nozzle Wire, Nozzle Cleaning Strainer, Nozzles, Nozzle Tool, Nozzle Wrench, Nozzle Brush and Nozzle Cleaning Tubes)
Ordering information: It includes user manual, nozzle cleaning solution, D & E empty bottle, Color 12 acessory kit, Barcode.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty