Would you like to log in again?
Pipette Stand, Rotary, Pipette Carousel 2, Eppendorf
Supplier: Eppendorf
3116000147 3116000139 3116000120
76204-696EA 80.8 USD InStock
76204-696 76204-692 76204-694
Pipette Stand, Rotary, Pipette Carousel 2, Eppendorf
Pipette Stands
These pipette carousels holds up to six manual single or multi channel pipettes.
  • Large carrying handle for easy transport of carousel from bench to bench
  • Large rubber feet protect stand from liquids spilled on lab bench
  • Small footprint with a diameter of 18.5 cm
  • Single pipette holders can be mounted to the carousel in two orientations turned by 180° to each other
  • Interchangeable pipette holders, no tools required
  • Pictograms clearly label pipette holders for easy assignment in the lab
These are without charging functionality.
Delivery information: Sticky tape included. Additional pipette holders are available separately.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty