Would you like to log in again?
Pipette Holder, Pipette Carousel 2, Eppendorf
Supplier: Eppendorf
3116000015 3116000023 3116000031 3116000058 3116000112
76204-690EA 52.4 USD InStock
76204-690 76204-776 76204-688 76204-778 76204-774
Pipette Holder, Pipette Carousel 2, Eppendorf
Pipette Stands
Carries up to six manual single or multi-channel Eppendorf pipettes.
  • Interchangeable and no tools required
  • Robust design that securely holds and protects pipettes
  • Handle for easy transport from bench to bench
Includes rotatable stand with completely new design that holds all current manual pipettes including Eppendorf Research®, Research® plus, Reference®, Reference® 2 or Biomaster® and most of their predecessors
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty