Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NucView® Caspase-3 Enzyme Substrates, Biotium

Supplier: Biotium

10408-T 10406 10406-T 10402-T 10402 10405-T 10407-T 10405 10403 10403-T 10408 10407
76204-726EA 80.25 USD InStock
76204-726 71003-844 75832-750 10154-178 71003-852 76204-798 10001-688 10154-180 76204-794 10001-596 75832-748 76204-796
NucView® Caspase-3 Enzyme Substrates, Biotium
Assays Cellular Assays
NucView® Caspase-3 Substrates are novel fluorescent probes that allow real-time detection of apoptosis by monitoring caspase-3/7 activity in intact cells using confocal microscopy, flow cytometry or microplate readers.

  • Real-time apoptosis monitoring: non-interfering with caspase activity unlike other caspase substrates or inhibitor-based assays
  • Bi-functional substrates: simultaneous detection of caspase 3/7 activity and visualization apoptotic nuclear morphology
  • Rapid, no-wash assay for microscopy or flow cytometry
  • Stable for several days for performing time-course assays
  • Tolerant of formaldehyde fixation and permeabilization
  • For use in adherent or suspension cell cultures
  • Storage at 4 °C

NucView® substrates consist of a fluorogenic DNA dye coupled to the caspase-3/7 DEVD recognition sequence. Upon entering apoptotic cells, the non-fluorescent substrate is cleaved by caspases-3/7 releasing a high-affinity DNA binding dye which migrates to the nucleus and becomes fluorescent upon binding to DNA.

NucView® 488 Caspase-3 Substrate, is also offered as a 1 mM solution in PBS (phosphate-buffered saline) for use in cells that are sensitive to DMSO toxicity. NucView® 405 and 530 are also available in 1 mM in PBS and trial sizes are available for both PBS and DMSO formulations. Note: In non-DMSO sensitive cell types, adding DMSO during the substrate incubation may enhance NucView staining.

Available in 3 color options:
NucView® 488: Ex/Em (bound to DNA) 500 nm/530 nm, NucView® 405 (bound to DNA): Ex/Em 429 nm/469 nm, NucView® 530 (bound to DNA): Ex/Em 528 nm/563 nm.

Certifications: NucView enzyme substrate technology is covered by U.S. Patent No. 8,092,784.

Ordering information: In addition to stand-alone NucView substrates, we offer a range of NucView® 488 live cell and apoptosis assay kits with detailed application protocols.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty