Would you like to log in again?
Brilliant III Ultra-Fast SYBR® Green ROX QPCR Master Mix, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Brilliant III
600889 600903 600903 600904 600892 600904 600892 600889
76204-816EA 483.64 USD InStock
76204-816 76204-848 CA76204-812 76204-812 76204-850 CA76204-850 CA76204-848 CA76204-816
Brilliant III Ultra-Fast SYBR® Green ROX QPCR Master Mix, Agilent Technologies
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
High performance, ultra-sensitive, SYBR® Green QPCR master mix reagent with ROX concentration for reliable quantification across a wide range of targets and templates.
  • Reliable and reproducible data
  • Novel fast Taq mutant for qPCR results in under 35 minutes Resistant to common QPCR inhibitors ( whole blood, humic acid, NaCl)
  • Enhanced rapid hot start capability saves time and reduces primer-dimer formation
  • Convenient pre-blended formulations compatible with any SYBR Green dye
Available with low or high ROX concentrations.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty