Would you like to log in again?
Brilliant III Ultra-Fast Probe ROX qPCR Master Mix, Agilent Technologies
Supplier: Agilent Technologies
Brilliant III
600898 600899 600899 600890 600898 600888 600890 600888
76204-814EA 483.64 USD InStock
76204-814 CA76204-814 76204-854 CA76204-854 CA76204-852 76204-852 76204-818 CA76204-818
Brilliant III Ultra-Fast Probe ROX qPCR Master Mix, Agilent Technologies
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
High performance, ultra-sensitive, probe QPCR master mix reagent with high or low ROX concentration for reliable quantification across a wide range of targets and templates.
  • Resistant to common QPCR inhibitors (whole blood, humic acid, NaCl)
  • Extremely fast while maintaining sensitivity
  • Optimized fast cycling formulation ensures reliable and reproducible data with shorter run times
Brilliant III Ultra-Fast maintains amplification efficiency, R2, dynamic range and detection sensitivity all while shortening the total cycling time by ~60% compared to standard cycling.
High performance, ultra-sensitive, probe QPCR master mix reagent with high/low ROX concentration for reliable quantification across a wide range of targets and templates. Convenient pre-blended formulations compatible with any sequence-specific probe detection chemistry.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty