Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Spin Concentrators, 50K MWCo, Agilent Technologies
Supplier: Agilent Technologies
5188-5202 5188-5202
76204-860PK 262.48 USD InStock
CA76204-860 76204-860
Spin Concentrators, 50K MWCo, Agilent Technologies
Filters Centrifugal Filters
Spin concentrators are devices used to concentrate protein samples via low-speed centrifugation through MWCo filters into a collection tube.
  • Contains a polyethersulfone (PES) membrane that is compatible with biological fluids and most aqueous solutions with pore sizes of 50 kDa
  • Active membrane area of 2.0 cm2 or 4.0 cm2
  • Durable and designed to minimize membrane fouling and allow fast concentration, even with particles in the solution
Designed to fit a standard 15 ml conical collection tube or a 50 ml collection tube (not provided with concentrator). Designed to be used in either swing-bucket or fixed angle rotor centrifuge equipment at maximum 5000 g or 2000 g.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty