Would you like to log in again?
Multiple Affinity Removal Spin Cartridge Reagent Kit, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5188-5249 5188-5252 5188-5254 5188-5254 5188-5252 5188-5253 5188-5253 5188-5250 5188-5251 5188-5251 5188-5249 5188-5250
76204-902PK 18.23 USD InStock
76204-902 CA76204-902 76204-904 CA76204-910 CA76204-900 76204-900 76204-910 CA76204-908 76204-906 CA76204-906 76204-908 CA76204-904
Multiple Affinity Removal Spin Cartridge Reagent Kit, Agilent Technologies
Protein Purification Protein Purification Kits
The affinity removal spin cartridge reagent kit is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
Delivery information: Kit includes Buffer A, 1 bottle (1 l) Buffer B, 1 bottle (1 l), Spin filters, 2 packs, Spin concentrators, 5 KDa MWCo (4 ml) 1 pack, Luer-Lok adapter 1 pack, Luer-Lok syringes (5 ml) 1 pack, Microtubes, 6 packs, Caps and plugs 1 pack, PTFE Luer-Lok needles 1 pack.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty