Would you like to log in again?
Multiple Affinity Removal Starter Reagent Kit, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5185-5989 5185-5987 5185-5986 5185-5988 5185-5990 5185-5987 5185-5990 5185-5991 5185-5988 5185-5989 5185-5986 5185-5991
76204-924EA 181.53 USD InStock
76204-924 CA76204-924 76204-926 CA76204-922 76204-920 CA76204-920 76204-922 CA76204-918 76204-928 CA76204-928 CA76204-926 76204-918
Multiple Affinity Removal Starter Reagent Kit, Agilent Technologies
Protein Purification Protein Purification Kits
Starter Reagent Kit for Multiple Affinity Removal LC Columns.
Delivery information: Includes: 2 bottles Buffer A (1 l), 1 bottle Buffer B (1 l), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty