Would you like to log in again?
Rotors for Centrifuges, 5910 Ri
Supplier: Eppendorf
5895170005 5895160000 5895155006 5895171001 5895181007 5895180000
76205-006EA 2908.16 USD InStock
76205-006 76204-996 76204-998 76205-000 76205-004 76205-002
Rotors for Centrifuges, 5910 Ri
Centrifuges Rotors
Eppendorf Rotors for Centrifuge 5910 Ri - precision in refrigerated centrifugation. Tailor your experiments with varied capacities and speeds. Accessorize with adaptable buckets and adaptors for efficient sample processing. Ideal for temperature-sensitive applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options available
Experience precision in refrigerated centrifugation with Eppendorf Rotors designed for centrifuge 5910 Ri. These rotors offer a range of capacities and speeds, providing flexibility for various sample processing needs. Accessorize with adaptable buckets and adaptors, enhancing the versatility of your centrifuge setup. Ideal for temperature-sensitive applications, the Centrifuge 5910 Ri is further optimized with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your refrigerated centrifugation experience.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty