Would you like to log in again?
LIGHTING SYSTEM LED OR F/25/33CF CHAMBER
Supplier: CARON PRODUCTS & SERVICE
LED (Orange) Lighting System For 25/33 CF chamber^ LED (Orange) Lighting System, For 25 and 33 CF chambers, Includes LED lighting strips located on the side walls, a 24 hour timer for day/night simulation, Not intended for plant growth, Narrow range (500-700nm), Factory installed at time of order
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LIGHTING SYSTEM LED OR F/25/33CF CHAMBER 76205-466 LGHT340 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty