Would you like to log in again?
LINER SPME ARROW 1.7MM SSL INJECTOR PK 3
Supplier: Thermo Scientific
SPME Arrow Liner ID 1.7mm^ SPME Arrow Liner ID 1.7mm, for SSL Injector Thermo Trace 1300, pk of 3 pcs, Only for SPME Arrow with OD 1.5mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LINER SPME ARROW 1.7MM SSL INJECTOR PK 3 76207-554 453A0415 Pack of 3 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty