Would you like to log in again?
VWR PAPER CR LF 22# GOLD 11X17 CS1250
Supplier: VWR International
VWR* Cleanroom paper 22 Gold 11x17in^ VWR* Cleanroom paper 22, Color: Gold, Latex free, They are ideal for documentation in critical environments, where very low particle and ESD generation is important, This clean room product is double bagged and Class 10 Certified, Dimension: 11x17in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR PAPER CR LF 22# GOLD 11X17 CS1250 76207-660 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty