Would you like to log in again?
Choice® SuperSorb Cleanroom-Laundered, Sealed-Edge Wiper, Berkshire
Supplier: Berkshire
Choice® SuperSorb
CHSS09.14 CHSS09B.14 CHSS12.12
89260-920CS 182.45 USD InStock
89260-920 76670-464 76207-696
Choice® SuperSorb Cleanroom-Laundered, Sealed-Edge Wiper, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Choice® SuperSorb is a two-ply, double knit, highly absorbent, ultrasonically sealed-edge, cleanroom-laundered wiper for reduced contamination. Made from 100% continuous filament knit polyester.
  • Critically low particles, fibers, ions, and extractables
  • High abrasion resistance
  • Chemically compatible with IPA, Acetone, and other solvents
  • Pinsonic design adds surface texture that aids in stubborn residue removal
Sealed edges increase durability and minimize fiber and particle release. Designed for the highest level of contamination control in critical processing applications, these wipers are excellent for the most critical spill control and cleaning applications.
Applications include sorbency for spill control in processing areas, stencil and print roll cleaning, cleaning of medical device products, applying and removing cleaning and disinfecting solutions. Steam autoclavable for aseptic environments.
Certifications: Complies with RoHS requirements.
Ordering information: Packaged in ISO class 4 (class 10) cleanroom.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty