Would you like to log in again?
MICROCHEM® AlphaTec® 600 Oversleeves, Ansell
Supplier: Ansell
Microchem® by AlphaTec®
GR40-W-92-600-00
76207-746CS 773.62 USD InStock
76207-746
MICROCHEM® AlphaTec® 600 Oversleeves, Ansell
Disposable Garments
Model 600 oversleeves are designed for use with the MICROCHEM® by AlphaTec® 4000 coveralls.
  • Multi-layer non woven barrier laminate fabric
  • Elasticated at both ends
  • Welded seams
Packaging: Sleeves are packaged 100 pairs per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty