Would you like to log in again?
Oakton® ORP Testr™ 50 Waterproof Pocket ORP Tester, Premium 50 Series, Cole-Parmer
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
35634-45
76208-094EA 497.97 USD InStock
76208-094
Oakton® ORP Testr™ 50 Waterproof Pocket ORP Tester, Premium 50 Series, Cole-Parmer
Electrochemical Meters pH and Multi-Parameter Meters
Ideal for checking sanitizing solutions, chromate reduction, cyanide oxidation, and other ORP readings.
  • Replaceable, double-junction electrode sensors save time and money
  • Easy-to-navigate interface for quick setup and calibration
  • Waterproof
  • LCD display
Redesigned leakproof cap functions as a sample cup, hydrating cup, and as a stabilizing base to stand meter upright.
Ordering information: Includes protective plastic storage case, lanyard, and 4 AAA batteries.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty