Would you like to log in again?
TransferTop™ Urine Collection and Transfer Container, Globe Scientific
Supplier: Globe Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3856TP 3841 3852 3856L
76308-970CS 404.78 USD InStock
76308-970 76208-580 76308-968 76479-380
TransferTop™ Urine Collection and Transfer Container, Globe Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
TransferTop™ Urine Collection and Direct Transfer Cups feature an integrated transfer mechanism for "closed system" transfer of urine samples directly from the cup to a vacuum sample tube.
  • Requires no open transferring of samples for reduced spills, improves lab hygiene, and limits exposure to dangerous samples
  • Reduces risk of contamination for improved test results
  • Directly transfers samples to a tube suitable for analysis and transport
  • Works with most vacuum urine tubes
  • Provides consistent sample transfer volumes when used with like vacuum tubes
  • Improves efficiencies and sample throughput when compared to traditional open systems
With the TransferTop™ System, there is no need for laboratory personnel to open the collection cup and pour the sample into a tube for processing. Instead, the safety label is peeled back, and the vacuum tube inserted for simple, safe, efficient, and reliable sample transfer.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty