Would you like to log in again?
Programmable Refrigerant Incubators, Yamato
Supplier: Yamato
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
IN-604115V IN-804W220V IN-604220V IN-604W115V IN-604WMK115V IN-604WMK220V IN-604W220V IN-804220V IN-804W115V IN-804115V
76209-618EA 6348.77 USD InStock
76209-618 76209-636 76209-626 76209-628 76209-632 76209-622 76209-634 76209-624 76209-630 76209-620
Programmable Refrigerant Incubators, Yamato
Incubators Cooled Incubators
Standard low temperature forced convection incubators applicable for low temperature tests and environmental tests.
  • Inner glass door keeps temperature stable during sample observation
  • Designed with a large dual glass door and inner door that forms a triplex glass door for improved heat retention (Models IN604W/804W)
  • Interior light for better sample visibility (Models IN604W/804W)
  • Optional slide shaker table available to put in and take out samples easily (Models IN604W/804W)
  • Digital PID Controller
It has an operating temperature range of –10 to+50 °C and has high accuracy temperature control and temperature distribution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty