Would you like to log in again?
ESD SMD Boxes, Transforming Technologies
Supplier: Transforming-Technologies
SM0873 SM0881 SM0883 SM0880 SM0882 SM0874 SM0876 SM0875
76209-948PK 279.61 USD InStock
76209-948 76209-956 76209-958 76210-004 76209-952 76209-954 76209-960 76209-950
ESD SMD Boxes, Transforming Technologies
Boxes ESD-Safe Boxes
Static-safe SMD boxes with spring-loaded hinges are designed to interlock for easy organization and storage.
  • Ideal for engineering labs
  • Available with conductive black or clear metalized lids
SMD plastic boxes have a unique interlocking modular design. Spring-loaded lids have a stay-open feature for convenient loading and removal of parts. Surface resistivity of 10² to 10⁵ ohms/cm². They are ideal for engineering labs, kitting, in-process handling, and safe transport of static-sensitive components.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty