Would you like to log in again?
ESD Safe SMD Component Storage Box Cabinet, Transforming Technologies
Supplier: Transforming-Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SM0810
76210-002EA 680.54 USD InStock
76210-002
ESD Safe SMD Component Storage Box Cabinet, Transforming Technologies
Boxes ESD-Safe Boxes
Organize, protect, and store interlocking SMD boxes in this six-drawer conductive cabinet.
  • Made of volume loaded conductive polystyrene
  • Tray compatible size combination
  • Spring loaded lid
  • Recyclable
The six-drawer cabinet allows for common storage of ESD safe boxes. Specifically compatible with the SM0880 and SM0870 series SMD boxes.
Certifications: Complies with EN 61340-5-1. Meets or exceeds requirements of ANSI ESD-S20.20
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty