Would you like to log in again?
WHEATON® 76-Series Vials™, DWK Life Sciences
Supplier: DWK Life Sciences
76-1236B
76210-034PK 100.11 USD InStock
76210-034
WHEATON® 76-Series Vials™, DWK Life Sciences
Vials
MicroLiter’s 76-Series Vials™ for Agilent autosamplers have a modified neck finish that assists in a repeatable seating of the consistent, gripper against the bottom of the cap.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty