Would you like to log in again?
Low Profile Common Point Ground Cords, Transforming Technologies
Supplier: Transforming-Technologies
CP2522 CP2524
76210-042EA 12.79 USD InStock
76210-042 76210-044
Low Profile Common Point Ground Cords, Transforming Technologies
Grounders Grounding Wires
Low profile common point ground cords connect multiple aspects of an ESD workstation to ground with a smaller connection point.
  • Common point ground cord for two wrist straps and ESD mat
  • One meg resistor for safety
Highly durable and reliable Low Profile Common Point Ground (CPG) Cords are equipped with two standard banana jacks, and will ground up to two operators at the same time. The low profile common point ground includes a 15’ long black PVC cord made with 7 strand tinsel wire to #10 ring terminal. The molded connection point houses a 1 megOhm 1/4 watt resistor and attaches to table mats via a 10 mm stud.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty