Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Savant™ SpeedVac™ Modular Vacuum Concentrators, Thermo Scientific
Supplier: Thermo Scientific
Savant™ SpeedVac™
SPD210P1-115 SPD210-115 SPD210P2-115
76210-400EA 9739.12 USD InStock
76210-400 76210-564 76210-568
Savant™ SpeedVac™ Modular Vacuum Concentrators, Thermo Scientific
Evaporators Centrifugal Evaporators
The Thermo Scientific™ Savant™ SpeedVac™ SPD210 vacuum concentrator is a large-capacity vacuum concentrator with a modular design, ideal for drying aqueous and organic solvents.
  • Kit
  • Medium capacity
  • Variety of rotors
  • Glass safety cover
  • PTFE coated aluminum chamber
The SpeedVac™ SPD210 is capable of drying larger quantities of samples at one time and will support your high-volume sample preparation needs.
Ordering information: Supplied as different packages, please see description for details of what is included in specific kits/packages.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty