Would you like to log in again?
LABEL SOLVENT RESISTANT RED 3X5 IN RL250
Supplier: GA INTERNATIONAL INC
Solvent Resistant Removable Color Label 3x5in^ Solvent Resistant Removable Color Label, Core: 1in, Color: red, Perforated, Adhesive: Removable, Label across: 1, For Containers, chemical-resistant, for thermal-transfer barcode printers, extra-durable, Labe
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LABEL SOLVENT RESISTANT RED 3X5 IN RL250 76210-454 807656033358 Roll of 250 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty