Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carboy Spill Containment Tray, Dynalon

Supplier: Dynalon

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

107364
76210-668EA 780.57 USD InStock
76210-668
Carboy Spill Containment Tray, Dynalon
Trays/Dishes/Pots Spillage Trays/Pans
Designed for laboratory carboys and other large containers with spigots to contain liquid spills.

  • Provides a safer way to dispense chemicals or other liquids in the lab
  • Protects cabinets, laboratory benches, and floors from liquid spills
  • Each tray contains two convenient drain ports and 0.5" tubulation connection on lower trough
  • Built-in lower bin for convenient storage of other labware or small accessories
  • 8" shelf height to accommodate up to a 5000mL beaker

Carboy spill trays are fabricated with 0.25" thick chemically resistant HDPE and welded construction featuring a 0.5" skid-resistant top shelf. The built-in side handles for convenient transport.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty