Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dippers, HDPE, Dynalon

Supplier: Dynalon

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

106525 106515 106505 107075 107095
76210-690EA 194.26 USD InStock
76210-690 76210-694 76210-692 76210-676 76210-696
Dippers, HDPE, Dynalon
Samplers Sample Dippers
High density polyethylene dipper for removing samples from large tanks.

  • Graduated plastic dipper
  • 2 pour spouts
  • PVC hand grip on 1" diameter HDPE rod
  • 500 mL and 1000 mL beaker welded to the rigid handle at a 45° angle

Made in the USA, these plastic dippers are lightweight but durable.

Packaging: The 12' size is supplied with a two-piece handle for economy in shipping and storage.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty