Would you like to log in again?
Liquid Sampler, HDPE, Dynalon
Supplier: Dynalon
114505-0500 114515-0500
76210-790EA 537.51 USD InStock
76210-790 76210-788
Liquid Sampler, HDPE, Dynalon
Samplers Fluid Samplers
This sampling device is perfect for corrosive environments and for hard to reach places.
  • Excellent for industrial and environmental applications
Plastic Liquid Sampler is manufactured from chemically resistant high density polyethylene.
The included bottles screw into the end of the handle and is filled via four 5mm diameter ports (2 allowing liquid to enter and 2 allowing air to escape). All rods are 1.25" O.D.
Packaging: Two-piece handle for UPS shipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty