Would you like to log in again?
Vydac® Chromatographic Silica, W.R. Grace
Supplier: W.R. Grace
Vydac®
5167350 5149682 3101618 3101615 3101621 3114821
76210-882EA 1061.58 USD InStock
76210-882 76210-880 76210-874 76210-872 76210-878 76210-876
Vydac® Chromatographic Silica, W.R. Grace
Chromatography Sorbents HPLC Sorbents
Vydac® chromatographic resin is excellent for biopurification. Proven in applications for protein, peptide, and oligonucliotide separations.
  • Completely scalable to ton quantities
  • High recovery of peptides and proteins
  • Available in a wide range of bonding chemistries
Grace pioneered one of the first platforms for reverse phase protein and peptide separation. Referenced in more than 9000 patents, Vydac® resin is the phase to which all others are compared. It is relied upon in numerous commercial scale protein purifications. High recovery of proteins makes this product more cost effetive than alternatives and is available in a wide range of bonding chemistries for alternate selectivities.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty