Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
DAVISIL® Wide Pore Chromatographic Silica, W.R. Grace
Supplier: W.R. Grace
5034754 5143586 5030057 5134301 5143587 5153368 5143590
76183-138EA 1725.14 USD InStock
76183-138 76183-126 76183-136 76183-134 76183-122 76183-132 76210-884
DAVISIL® Wide Pore Chromatographic Silica, W.R. Grace
Chromatography Sorbents HPLC Sorbents
Unique Wide Pore and Extra-Wide Pore silicas are a cost effective solution for purification of large molecules.
  • Pore Sizes 250 – 2500 Å
  • Exceptional mechanical strength
  • Large pore sizes with narrow pore size distribution
Grace Davisil® Wide Pore Chromatographic Silica is suitable for use as a packing for liquid chromatography normal phase separations which require pore sizes from 250 – 4500 Å. Applications include separation and purification of Biomolecules such as large peptides, oligonucleotides, proteins, vaccines, virus-like particles, RNA and DNA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty