Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
DAVISIL® 150Å Chromatographic Silica, W.R. Grace
Supplier: W.R. Grace
5076059 5173139 5183333 5183331 5173140 5037727 5057993
76183-128EA 4078.73 USD InStock
76183-128 76183-178 76210-928 76183-120 76210-930 76183-180 76183-118
DAVISIL® 150Å Chromatographic Silica, W.R. Grace
Chromatography Sorbents HPLC Sorbents
Proven performance and high quality chromatographic silica.
  • Proven performance
  • Predicable scale-up
  • Exceptional quality and consistency
  • Wide selection
Grace Davisil® 150 Å Chromatographic Silica is suitable for use as a packing for liquid chromatography normal phase separations. Applications include separation and purification of small molecule pharmaceuticals, vitamins, natural products, and small peptides. Scalable from analytical to process scale.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty