Would you like to log in again?
Direct-zol™ RNA Kits, Zymo Research
Supplier: Zymo Research
Direct-zol™
R2055 R2053 R2072 R2056 R2062 R2071 R2102 R2050 R2073 R2101 R2052 R2103 R2070 R2057 R2061 R2054 R2070T R2100 R2063 R2051 R2060
76211-350KT 1595.3 USD InStock
76211-350 76211-340 76211-330 76211-342 76211-332 76211-344 76211-334 76197-150 76211-324 76211-346 76211-348 76211-326 76211-328 76020-108 76020-648 76020-110 76020-452 76020-642 76020-456 76020-112 76020-454
Direct-zol™ RNA Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
RNA from TRIzol® in seven minutes.
  • Extract RNA from any sample (cells, tissues, blood, biological liquids, etc.)
  • Easy Handling: No phase separation or precipitation steps
  • NGS-ready: Ultra-pure RNA without phenol carryover, no DNA contamination (DNase I included)
  • Total RNA Recovery: All RNA >17 nt is recovered
The Direct-zol™ RNA Kits provide a streamlined method for the purification of up to 100 µg (per prep) of high-quality RNA directly from samples in TRI Reagent® or similar. Total RNA, including small RNAs (17 to 200 nt), is effectively isolated from a variety of sample sources (cells, tissues, serum, plasma, blood, biological liquids, etc.).
The procedure is easy. Simply apply a prepared sample in TRI Reagent® directly to the Zymo-Spin™ column and then bind, wash, and elute the RNA. No phase separation, precipitation, or post-purification steps are necessary. The eluted RNA is high quality and suitable for subsequent molecular manipulation and analysis (including RT-PCR, transcription profiling, hybridization, sequencing and more).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty