Would you like to log in again?
Dihydrofolate Reductase Activity Kit (Colorimetric), BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB239705-100TEST
76211-360EA 858.31 USD InStock
76211-360
Dihydrofolate Reductase Activity Kit (Colorimetric), BioVision
Assays Enzyme Assays
The assay is simple, sensitive and can detect as low as 4 mU/ml in a variety of samples.
  • Rapid, sensitive and convenient
  • The kit measures total Activity of DHFR Activity with limit of quantification 4 mU/ml using recombinant DHFR
Dihydrofolate Reductase (DHFR; 5,6,7,8-tetrahydrofolate NADP oxidoreductase; EC 1.5.1.3), is a ubiquitous enzyme that is present in all eukaryotic and prokaryotic cells. It catalyzes the reduction of dihydrofolate (FH2) to tetrahydrofolate (FH4) using NADPH as a cofactor. FH4 is essential for a number of enzymes that are necessary for the de novo synthesis of purines, thymidylic acid and some amino acids. Inactivation of the DHFR enzymatic activity causes reduction of the intracellular level of FH4, inhibition of RNA and DNA synthesis, and cell death. For this reason, DHFR has been a critically important enzyme as a molecular target in drug discovery. Biovision’s Dihydrofolate Reductase assay kit is based on the ability of DHFR to catalyze the oxidation of NADPH. The reaction progress is followed by monitoring the decrease in absorbance at 340 nm. Our assay has been optimized to be carried out in a 96- well plate. The assay is simple, sensitive and can detect as low as 4 mU/ml in a variety of samples.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty