Would you like to log in again?
QuickDetect™ CTLA4 (Human) ELISA Kit, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
QuickDetect™
AB285255-96TEST
76211-686EA 1654.77 USD InStock
76211-686
QuickDetect™ CTLA4 (Human) ELISA Kit, BioVision
Assays ELISAs
A rapid sandwich ELISA kit to quantify CTLA4 in human serum, plasma, cell culture supernatnat
  • Easy, convenient and time-saving method to measure the level of CTLA4 in human serum, plasma, culture supernatants and other biological fluids.
  • The entire assay can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results.
  • Detection Range: 3.7 - 120 ng/ml
Inhibitory receptor acting as a major negative regulator of T-cell responses. The affinity of CTLA4 for its natural B7 family ligands, CD80 and CD86, is considerably stronger than the affinity of their cognate stimulatory coreceptor CD28. BioVision’s QuickDetect CTLA4 ELISA kit is a sandwich ELISA assay for measuring the quantity of human CTLA4 in serum, plasma and cell culture and other biological fluid. The entire process can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty