Would you like to log in again?
6-Phosphogluconate Dehydrogenase Activity Colorimetric Assay Kit, BioVision, Inc.
Supplier: BioVision
AB241016-100TEST
76211-842EA 1206.62 USD InStock
76211-842
6-Phosphogluconate Dehydrogenase Activity Colorimetric Assay Kit, BioVision, Inc.
Assays Enzyme Assays
Kit provides quick, sensitive & easy way for measuring 6-PGD activity in various biological samples
  • Simple, rapid & convenient assay to measure 6-PGD Activity in biological samples
  • Includes Positive Control
6-Phosphogluconate dehydrogenase (6-PGD, EC 1.1.1.44) is an enzyme in the pentose phosphate pathway that catalyzes the oxidative decarboxylation of 6-phosphogluconate to ribulose-5-phosphate with simultaneous reduction of NADP+ to NADPH. Ribulose-5-phosphate is used for production of nucleotides and nucleic acids and can also be reversibly converted to glyceraldehyde-3-phosphate and fructose-6-phosphate which can be used in the glycolysis pathway. It also has a vital role in production of NADPH, which is utilized as a reducing agent in numerous biosynthetic pathways and is important in prevention of oxidative damage. In humans, 6-PGD deficiency is an autosomal hereditary disorder that can result in depletion of NADPH in red blood cells. As NADPH is required for regeneration of the cellular antioxidant glutathione in red blood cells, 6-PGD deficiency increases the risk of hemolytic anemia in situations of oxidative stress. On the other hand, inhibition of 6-PGD has been linked to anti-cancer activity by decreasing RNA biosynthesis and increasing the buildup of reactive oxygen species in tumor cells. BioVision’s 6-PGD assay kit provides a quick, sensitive and easy way for measuring 6-PGD activity in various biological samples. In this assay, 6-PGD converts 6-phosphogluconate into an intermediate, generating NADPH, which subsequently reduces a colorless probe into a strongly colored product detectable by absorbance at 460 nm. The assay is high-throughput adaptable and can detect less than 0.05 mU of 6-PGD activity
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty