Would you like to log in again?
Cystathionine β Synthase Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB283406-100TEST
76211-844EA 1330.27 USD InStock
76211-844
Cystathionine β Synthase Inhibitor Screening Kit (Fluorometric), BioVision
Assays Enzyme Assays
Enables customers to screen inhibitors of CβS and quantify their therapeutic potential.
  • Simple and High throughput adaptable
  • Fluorometric Detection
  • Screen within 60 min
Cystathionine β Synthase (EC 4.2.1.22, CβS) is a Pyridoxal 5-Phosphate-dependent enzyme that uses cysteine and homocysteine as substrates to catalyze the formation of H2S and cystathionine. CβS is well-known for its role in human sulfur metabolism. The overexpression of CβS has been implicated in Down Syndrome, and also results in homocystinuria; or low levels of homocysteine in blood. Homocystinuria leads to a mutli-systemic disorder of the connective tissue, muscles, and central nervous system (CNS). For that reason, pharmacological inhibitors of human CβS present attractive therapeutic potential in order to restore baseline CβS activity and blood homocysteine levels. It has been shown that aminooxyacetic acid (AOAA) is an irreversible inhibitor that targets the PLP-binding site of CβS. That interaction inhibits the transaminase reaction that would yield cystathionine and H2S in wildtype conditions. BioVision’s Cystathionine β Synthase Inhibitor Screening Kit is the first available kit that enables customers to screen inhibitors of CβS and quantify their therapeutic potential. The kit has a simple, easy-to-follow protocol and is high-throughput adaptable. In this kit, the product hydrogen sulfide reacts with the non-fluorescent azido-functional group to yield a fluorescent amino group (Ex/Em = 368/460 nm). In the presence of AOAA, CβS activity is inhibited, which subsequently abolishes the production of H2S and thus the fluorescent signal is reduced.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty