Would you like to log in again?
EZCell™ Phagocytosis Assay Kit (Red E. coli), BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB235901-100TEST
76211-926EA 712.47 USD InStock
76211-926
EZCell™ Phagocytosis Assay Kit (Red E. coli), BioVision
Assays Cellular Assays
Kit provides a robust screening system for activators & inhibitors of phagocytosis & TLR ligands
  • Simple & Rapid Protocol
  • Convenient: Non-Radioactive, no special handling or disposal required
  • High-Throughput
  • Accurate: reproducible results with low intra & inter assay variability
Phagocytosis in mammals serves as an important first line defense mechanism against invading pathogens. It is also essential for continuous clearance of dying cells, tissue remodeling, and acquisition of nutrients for some cells. Phagocytosis is a specific form of endocytosis initiated by recognition and binding of foreign particles by cell surface receptors, followed by their engulfment, and formation of phagosomes. Maturing phagosomes transform to phagolysosomes which destroy the pathogen through enzymes and toxic peroxides. E. coli and other bacterial strains are often used as a pathogen in phagocytosis assays. BioVision’s EZCell™ Phagocytosis Assay Kit (Red E. coli) utilizes heat-killed, fluorescently pre-labeled E. coli particles as a tool for rapid and accurate detection and quantification of in vitro phagocytosis by fluorescent microscope, spectrophotometer or flow cytometry. The kit provides a robust screening system for activators and/or inhibitors of phagocytosis and Toll-like receptors (TLR) ligands
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty