Would you like to log in again?
Mutant Isocitrate Dehydrogenase (Mutant IDH) Activity Assay Kit (Colorimetric), BioVision, Inc.
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB284568-100TEST
76211-958EA 1138.99 USD InStock
76211-958
Mutant Isocitrate Dehydrogenase (Mutant IDH) Activity Assay Kit (Colorimetric), BioVision, Inc.
Assays Enzyme Assays
First in vitro assay with a simple, sensitive, non-radiometric protocol for detection of Mutant IDH.
  • Simple procedure
  • Fast and convenient
In eukaryotic cells, Isocitrate Dehydrogenase (IDH1, IDH2 and IDH3) is an enzyme that catalyzes the decarboxylation of Isocitrate producing α-Ketoglutarate and CO2. Mutations in both isoforms of IDH (IDH1 and IDH2) are commonly found in human cancers. The Mutant Isocitrate Dehydrogenase (Mutant IDH) causes a “gain-of-function”, which reduces its affinity for isocitrate and favors the conversion of α-ketoglutarate to D-2-Hydroxyglutarate. D-2-Hydroxyglutarate (D2HG) is present at low level in normal cells and tissues, but is significantly elevated in metabolic diseases and various cancers. D2HG functions as an “oncometabolite” promoting cellular transformation. Recent studies show that increased Mutant IDH activity is associated with various cancers; therefore, detection of Mutant IDH activity is important for diagnosis and developing therapeutic strategies (e.g. Mutant IDH inhibitors). BioVision’s Mutant IDH assay kit provides a quick and simple assay to monitor the Mutant IDH activity in biological samples. In the assay, Mutant IDH oxidizes NADPH into an NADP+, which decreases the absorbance at 340 nm. The assay is simple, sensitive, and can detect Mutant Isocitrate Dehydrogenase activity lower than 2 mU/ml in a variety of cancer samples
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty