Would you like to log in again?
Turbo Toe Safety Steel Toe Caps, Magid®
Supplier: Magid
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Turbo Toe
TTXL TTM TTXS TTXXXL TTXXL TTXXS TTL TTS
76213-984PR 57.94 USD InStock
76213-984 76214-002 76213-986 76214-006 76214-004 76213-982 76214-008 76213-988
Turbo Toe Safety Steel Toe Caps, Magid®
Shoes and Boots
The Turbo Toe Steel Toe Cap overshoes protect the toe area from accidental impact, crushing or stubbing. It can be comfortably worn over low-heel, dress and running shoes and most boots.
  • Made of flexible, pliable PVC
  • Wide size range to accommodate both men and women
  • Toe is color-coded to indicate size
  • 100% waterproof, will not harden, stiff or crack
Treads under sole to prevent slipping and help move liquid away from the foot. Ideal for any working environment that carries a risk of toe damage or has a steel toe cap requirement. Perfect for visitors, casual or temporary workers, management, clerical staff and sales people.
Certifications: CE marked. Certified to standard Z334-14 – Protective Footwear. Class 7231 02 – Occupational Health & Safety Products – Grade 1: over the shoe toe protectors withstands impact velocity minimum of 3.32m/s. Tested to ASTM Standard F2413/F2412 and meets 75 lb. impact (I/75) and compression (C/75) resistance rating.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty