Would you like to log in again?
Histosette® II Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Supplier: Simport
Histosette® II
M492-8T M492-3T M492-7T M492-8T M492-10T M492-2T M492-6T M492-5T M492-10T M492-4T M492-3T M492-4T M492-12T M492-5T M492-2T M492-9T M492-11T M492-7T M492-12T M492-9T M492-11T M492-6T
76214-078CS 861.48 USD InStock
CA76214-076 CA76214-186 76214-078 76214-068 CA76214-184 76214-076 CA76214-074 76214-186 CA76214-078 CA76214-068 76214-070 76214-180 CA76214-080 76214-080 CA76214-182 76214-184 76214-074 CA76214-072 CA76214-070 CA76214-180 76214-072 76214-182
Histosette® II Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Embedding Cassettes Tissue Processing Cassettes
Secure specimens during processing with the Histosette® II Tissue Processing/Embedding Cassette.
  • In Quickload™ stacks
  • 45° anterior writing area
  • Open and close as often as necessary; cassettes will always relock securely
  • Back-mounted tab allowing an unobstructed anterior marking area
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Keep specimens safely submerged in liquid. The flow-through slots maximize fluid exchange. The one-piece integral lid eliminates the need for separate steel lids.
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty