Would you like to log in again?
Unisette™ Biopsy Processing Cassette in Quickload™ Stacks for Primera Printers, Simport Scientific
Supplier: Simport
Unisette™
M406-2T M406-7T M406-12T M406-4T M406-12T M406-6T M406-4T M406-3T M406-7T M406-9T M406-2T M406-6T M406-9T M406-11T M406-8T M406-5T M406-10T M406-10T M406-3T M406-8T M406-11T M406-5T
76214-364CS 398.17 USD InStock
CA76214-362 76214-364 CA76214-352 76214-354 CA76214-030 76214-362 CA76214-360 76214-352 CA76214-350 CA76214-366 76214-358 CA76214-356 76214-366 CA76214-364 76214-356 CA76214-354 76214-030 76214-360 76214-350 CA76214-348 76214-348 CA76214-358
Unisette™ Biopsy Processing Cassette in Quickload™ Stacks for Primera Printers, Simport Scientific
Embedding Cassettes Biopsy Processing Cassettes
Made of acetal and resistant to the chemical action of most histological solvents and suitable for automated printers.
  • In Quickload™ stacks, for Primera printers
  • 35° anterior writing area
  • One-piece construction with hinge-lock design prevents early separation of base and lid
  • Snap latch and hinge-lock design; cassettes will always relock securely
  • Allows for one-hand operation
Secure specimens during processing with the Unisette™ Biopsy Processing/Embedding Cassette. The snap latch and hinge-lock design allows for one-hand operation and always relocks securely without danger of specimen loss.
Certifications: CE-IVD.
Packaging: Each case contains 25 Quickload™ stacks of 40 cassettes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty