Would you like to log in again?
VWR® TraceableGO™ Bluetooth Datalogging Temperature and Humidity Meter
Supplier: Avantor
61010-43
76214-378EA 116.86 USD InStock
76214-378
VWR® TraceableGO™ Bluetooth Datalogging Temperature and Humidity Meter
Data Loggers
A Bluetooth device is the ideal transit partner for monitoring materials between destinations. Safely transport reagents, clinical samples, vaccines, pharmaceuticals, and food items to any location.
  • Press START to begin trip monitoring
  • Place device inside a small cooler or even an entire pallet of material
  • Once the trip has ended, double-press START/STOP to enable Bluetooth and export data using the TraceableGO™ mobile app
  • TraceableGO™ mobile app immediately identifies if the contents experienced conditions outside user-defined parameters
  • Compatible with TraceableGO or TraceableLIVE mobile apps
  • Simple, single button use
  • Dimensions:3.5×2×0.79"
With the TraceableGO™ mobile app, view if an alarm was triggered, and generate an instant data report. Device stores up to 90 alarm events, records readings once every 5 minutes by default, or the user may adjust the reading interval between one minute to once every 12 hours. Logging duration is 227 days (7.5 months). TraceableGO™ Bluetooth devices are pre-programmed with alarm values.
Bluetooth connection works with any Bluetooth-enabled devices. Once TraceableGO™ arrives, enable the Bluetooth transfer to a smartphone by opening the TraceableGO™ app (available on iTunes or Google Play) and pairing the device to a smartphone. Download the recorded data once the device is enabled to send a report directly from a mobile device.
Bluetooth hygrometer datalogger fills in the unknowns between destinations. Critical information(humidity and temperature, trip duration, MIN/MAX, battery level and alarms) all displayed on LCD at all times.
Certifications: CE marked. RoHS compliant. Traceable to NIST which assures accuracy from an ISO/IEC 17025:2017 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM mutual recognition agreement.
Accessories information: Data can be downloaded and devices reconfigured using the free TraceableGO™ app. Optional service allows an unlimited amount of stored data from an unlimited number of TraceableGO™ devices on a TraceableLIVE® account for $30/month.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty