Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® Traceable® Giant-Digits™ Radio Atomic Clock with Humidity

Supplier: VWR
VWR® Traceable®
90027-83
76214-380EA 179.3 USD InStock
76214-380
VWR® Traceable® Giant-Digits™ Radio Atomic Clock with Humidity
Timers Wall Clocks
An abdundance of features combine to create the perfect laboratory clock. View temperature and humidity readings from anywhere in the lab.

  • Giant-Digits™ 4.75" display is viewable from 200 feet
  • Simultaneously displays both F/C temperatures and relative humidity
  • Temperature range: 0 to 45 °C (32 to 113 °F); Resolution: 0.1°; Accuracy: ±1 °C
  • Humidity Range: 20 to 90% RH; Resolution: 1% RH; Accuracy: ±5% mid-range (±7% otherwise)
  • Accuracy is ¹/₁₀₀₀₀ of a second; Maintains better than 1-second accuracy for 25000 years
  • 1 second resolution

Unit features a radio signal from NIST’s Colorado atomic clock to automatically set correct date/time. A calendar displays month/date, and day. Additional features include alarm, temperature and humidity display, and desk and wall mount supplies.

Individually numbered Traceable® Certificate assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (national Institute of Standards and Technology).

Certifications: CE marked. RoHS compliant. Traceable to NIST for accuracy.

Delivery information: Batteries included.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty